biOrb® Air Terrarium Parts
Supplier: GEOGLOBAL PARTNERS
45550 46149 46151 46155
470234-736CS 328.2 CAD
470234-736 470235-096 470235-098 470235-100 470235-102 470235-104 470235-106
biOrb® Air Terrarium Parts
Animal Supplies Animal Environmental Enrichment
Don't Forget Your Maintenance Parts.
  • Specifically for biOrb® air
The Coir Compost is a treated and sterilized growth medium ideal for cultivating plant roots in the biOrb® AIR. The compost is made from cocopeat which is a renewable resource. One block is enough for a single biOrb® AIR.
The Misting Unit is capable of producing a misting volume of 200ml/h. The biOrb® AIR Misting Unit is designed to be used with biOrb® Humidimist for proper performance. The green capillary matting is made from PP, PET and viscose it is a traditional capillary mat combining good capillary action and plenty of water storage capacity for sub-irrigation.
The air filter cartridge is an open cell reticulated polyether based polyurethane foam, which has been post treated with activated carbon. No handling of activated carbon is needed offering a mess free experience. It is recommended to change the air filter cartridge every six months for optimum performance.
The Humidimist pack includes specially formulated remineralized water with low conductivity and reduced minerals to prevent precipitation in your biOrb® AIR. Includes four, 16.9 fl. oz. bottles, three with cap tops and one with a spray top. This reliable 24 V transformer rarely needs replacing but is very simple to install. The power source for the biOrb® AIR, and can not be used with other biOrb® products.