IBI Isolate DNA/RNA Reagent Kit, IBI Scientific
Supplier: IBI Scientific
IBI Isolate is a phenol, chloroform, and guanidine isothiocyanate based scalable solution for extracting high-quality total RNA as well as simultaneous extraction of RNA, DNA, and protein from a wide variety of samples.
- Sample Size: Scalable for blood, buffy coat, serum, plasma cultured cells and tissue
- Operation time: up to 60 minutes
- Can simultaneously extract DNA – RNA and Protein from the same sample
IBI Isolate is a phenol, chloroform, and guanidine isothiocyanate based scalable solution for extracting high-quality total RNA as well as simultaneous extraction of RNA, DNA, and protein from a wide variety of samples such as blood, buffy coat, plasma, serum, cultured cells, and tissue.
Certifications: For research and development purposes only.