PolarSafe™ Stainless Steel Freezer Racks, Argos Technologies
Supplier: Antyila Scientific
04399-40 04400-83 04406-79 04399-74 04400-53 04401-05 04400-37
76076-800EA 312.64 CAD
76076-800 76076-802 76076-804 76076-806 76077-108 76077-110 76077-112
PolarSafe™ Stainless Steel Freezer Racks, Argos Technologies
Racks Cryogenic Box Racks
PolarSafe™ Metal Freezer Storage Racks are constructed of high-quality, corrosion-resistant stainless steel.
  • Handles at both ends accommodate freezers with right- or left-hand door openings
  • Configured to fit all major ultra low (–80°) freezers on the market
Ordering information: RDP75A and RTB74A include locking rods.