Magid® Comfort Flex® Neoprene/Latex Chemical Gloves, Magid Glove
Supplier: Magid
Comfort Flex®
738-7 7388 73810 7389
76193-360DZ 66.01 CAD
76193-360 69100-566 69100-564 69100-568
Magid® Comfort Flex® Neoprene/Latex Chemical Gloves, Magid Glove
Gloves
The Comfort Flex® flock lined neoprene/latex blend waterproof glove provides superior chemical and mechanical resistance.
  • Flock/lined
  • Blue neoprene over yellow latex glove
  • Embossed finish for fantastic grip
  • Perfect for work with most chemicals
This liquid-proof glove features the unparalleled grip of an embossed diamond finish, and the chemical and abrasive resistance of neoprene and latex rubber.
Versatile and form-fitting, it is perfect for use in the aircraft industry, chemical processing, food processing, cleaning parts with MEK, fiberglass lay-up, and general maintenance.
Certifications: USDA accepted.
Caution: Products containing natural rubber latex may cause allergic reactions in some individuals.