DistriTip® Sterilized Syringe Tips for DISTRIMAN® Repetitive Pipettor, Gilson®
Supplier: GILSON, INC.
F164130 F164140 F164150
76178-256PK 251.83 CAD
76178-256 76178-258 76178-260
DistriTip® Sterilized Syringe Tips for DISTRIMAN® Repetitive Pipettor, Gilson®
The best quality to guarantee repetitive distribution results.
DistriTip® tips are polypropylene syringes with easy-to-read level indicators. Three models cover the entire volume range from 1 µl to 12.5 ml. More than 100 aliquots can be dispensed from each syringe. These sterilized syringe tips are individually wrapped and certified free of RNase, DNase, DNA, RNA, and protease. Used with the DISTRIMAN® positive-displacement repetitive pipettor, DistriTip® tips provide more accurate and precise measurements for viscous, dense, volatile, or hot liquids.
DistriTip® Syringe Tips are specifically designed for use with the DISTRIMAN® Repetitive Pipettor.