Conductive Thread
Supplier: BROWN DOG GADGETS
CP2031-01PK
470234-668EA 5.2 CAD
470234-668
Conductive Thread
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Re-imagine wires and the purpose of sewing with this clever material!
  • Steel/nylon mixture sews like thread
  • Resistance of 1 ohm per foot
  • 50 m (164') per spool
Conductive thread is included in many of our crazy circuits kits and is great for soft circuits and wearable projects, such as light-up bracelets, t-shirts, ties and sock puppets. It sews just like normal thread but is electrically conductive. Now, your sewn connections can also be circuit pathways. The possibilities are endless!
Recommended for hand stitching and hand washing only.