Nylon Conductive Tapes
Supplier: BROWN DOG GADGETS
K2029-05M P2030-20M
470234-664EA 8.7 CAD
470234-664 470234-666
Nylon Conductive Tapes
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Introducing: Maker Tape!
  • Blend of nylon and steel
  • Low resistance per foot
  • Strong and flexible, tear resistant
  • Overlapping layers make conducting electrical connection
Our favorite nylon-steel blend is now thinner and z-axis conductive. Just overlap two pieces of Maker Tape to create a solid electrical connection. This tape is much stronger and easier to work with than commonly used copper foil tapes. The ⅛" width tape is perfect for Crazy Circuits™ projects on LEGO™ bricks or compatible systems. The ¼" width is perfect for sticking down LEDs in PaperMore Product Information