ESF AF™ Insect Cell Culture Medium
Supplier: EXPRESSION SYSTEMS LLC
99-300-01 99-300-10
77705-808EA 142.6 CAD
77705-808 77705-810
ESF AF™ Insect Cell Culture Medium
Cell Culture Media
ESF AF™ insect cell culture medium is a complete serum-free, protein-free medium developed for robust cell growth, protein expression and baculovirus vector production for a wide range of insect cells.
  • Animal component-free (100%)
  • Comparable to ESF 921 medium for robust cell growth and expression
  • Sf9 cell densities of 20×106 cells/ml or greater
  • Ideal for scale-up manufacturing
  • Works with a wide variety of baculovirus and host cell platforms
  • Optimized for applications such as production of sub-unit vaccines, virus-like particles and gene therapy vectors (rAAV - all serotypes)
  • Complete, ready-to-use, contains L-Glutamine
  • Adapted cells available in suspension
ESF AF is an animal origin-free formulation for insect cell culture. It has been thoroughly tested on Sf9, Sf21, Tni (High Five™) and S2 cell lines and is comparable to ESF 921 for growth and expression. ESF AF is the logical choice for further manufacturing under cGMP/FDA regulatory compliance.
Ordering information: Available in 1 L bottles, 10 L bags, 20 L bags and 50 L drums.