BestBac Baculovirus CoTransfection Kit
Supplier: EXPRESSION SYSTEMS LLC
91-100 91-200
77705-742EA 1167.78 CAD
77705-742 77705-744
BestBac Baculovirus CoTransfection Kit
Transfection Reagents
BestBac Linearized Baculovirus DNA backbone has been engineered to deliver highly efficient recombination with all polyhedron gene locus based transfer vectors.
  • Unmatched productivity and stability for BEVS
  • Improved production methodology can reduce need for plaque purification
  • Produces high titers of 5 E8 to 3 E9 IU/ml of recombinant baculovirus in P1 stock
  • Generates P1 stock as quickly as Bac-to-Bac® and other systems
  • Each BestBac vial contains enough material for a minimum of five cotransfections
  • Suitable for other expression systems including BacMam™
The resulting virus vector provides superior stability and purity. BestBac is available with a deletion or wild-type at the v-cath/chiA region.
Delivery information: Includes: BestBac DNA, pVL1393-lacZ control vector, Transfection medium, Expres² TR transfection reagent. Priority overnight.