PIPETMAN® Diamond Tips, BLISTER Refill, Gilson®
Supplier: GILSON, INC.
F172403 F172503 F172703 F172303 F172203 F172200 F172201 F172510 F172511 F172310 F172311 F172613 F172513 F172313 F172213 F172500 F172501 F172300 F172210 F172211 F172301 F172400 F172401 F172603 F172800 F172801 F172803
76178-276PK 318.21 CAD
76178-276 76178-278 76178-280 76178-282 76178-284 76178-286 76178-288 76178-290 76178-292 76178-294 76178-296 76178-298 76178-302 76178-304 76178-306 76178-592 76178-594 76178-596 76180-028 76180-030 76180-354 76180-356 76180-358 76180-360 76470-104 76470-106 76470-108
PIPETMAN® Diamond Tips, BLISTER Refill, Gilson®
Pipette Tips
PIPETMAN® DIAMOND Tips are designed to fit your PIPETMAN and PIPETMAX® perfectly. They deliver accurate and precise pipetting when used in conjunction with Gilson pipettes.
  • Individually wrapped racks of tips that are convenient, clean, and eco-friendly
  • Ready to use, just peel the lid and insert into the empty box
  • Uses 45% less plastic than regular rack box
  • Fully autoclavable, available as sterilized and filter tips
Smart way to fefill:
BLISTER REFILLS contain individually wrapped racks of tips that are convenient, clean, and eco-friendly. The racks are fully autoclavable and are color-coded for easy identification and compliance with Good Laboratory Practice (GLP). BLISTER REFILLS are available as autoclavable, sterilized, or sterilized filter tips. PIPETMAN DIAMOND filter tips contain a high-density polyethylene filter to protect against aerosol-born contamination and do not contain any sealing additives that can be released into your sample so you can be confident in your results.
Accurate and precise:
Using PIPETMAN DIAMOND Tips ensures maximum accuracy and precision and delivers consistent results. The perfect air-tight seal prevents leaks. The tips are made of pure polypropylene to deliver the correct volume and ensure complete dispensing. Volume graduations make it easy to check the amount aspirated visually.
Convenient:
Ready to use; peel the lid and insert it into an empty box.
Gilson tips are manufactured in a cleanroom environment, ensuring they are free of detectable contaminants that can affect your results. From mold to bench, our tips are certified and traceable to meet the most stringent demands in many applications.