Transfection Medium
Supplier: EXPRESSION SYSTEMS LLC
95-020-020 95-020-100
77705-784EA 135.09 CAD
77705-784 77705-786
Transfection Medium
Transfection Reagents
Transfection medium is designed to complement ESF 921 and ESF AF insect cell culture media for the co-transfection or transfection step in baculovirus vector production.
  • Serum-free, animal-free formulation works seamlessly with ESF 921 and ESF AF
  • Appropriate for PEI or lipofection mediated transfection
  • Optimized for use with Expres2 TR Transfection Reagent
  • Increases virus vector production and transient expression
  • Available in 20 or 100 ml bottles
It facilitates vector DNA uptake by the insect host cells.
Delivery information: Standard overnight.