PIPETMAN® Diamond Tips, TowerPack™, Gilson®
Supplier: GILSON, INC.
F167100 F167101 F167102 F167103 F167105 F167104 F167201 F167202 F167203 F167205 F167204
76178-228EA 27.41 CAD
76178-228 76178-230 76178-232 76178-234 76178-236 76178-238 76178-240 76178-242 76178-244 76178-246 76178-248
PIPETMAN® Diamond Tips, TowerPack™, Gilson®
Pipette Tips
PIPETMAN® DIAMOND Tips are designed to fit your PIPETMAN perfectly and deliver accurate and precise pipetting when used in conjunction with Gilson pipettes. The TOWERPACK™ refill system allows you to refill tips quickly and easily. The tip racks are color-coded for easy identification and compliance with Good Laboratory Practice (GLP).
  • Accurate and precise
  • Reliable
  • Convenient use and storage
Using PIPETMAN DIAMOND Tips with your PIPETMAN ensures maximum accuracy and precision and delivers consistent results. The perfect air-tight seal prevents leaks.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable