Streptavidin
Supplier: Biolegend
405210
CA405210-BLEA
205.86
CAD
CA405210-BL
Streptavidin
Proteins and Peptides
Bilolegend® streptavidin-HRP is a peroxidase-conjugated streptavidin biotin-binding protein with high affinity for detecting biotinylated antibodies in ELISA, ELISPOT, IHC or Western blotting.
- Reactivity: Mouse, Human, Rat
- Storage: 2 to 8 °C
Streptavidin-HRP is used as a second step reagent for indirect enzymatic labelling, in conjunction with biotinylated primary antibodies.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...