Water, high purity for histology

Supplier: AZER SCIENTIFIC INC

Synonyms: Hydrogen oxide

ES611 ES612
89176-814CS 140.81 CAD
89176-814 89176-816
Water, high purity for histology
Water

High Purity Water is recommended for improved slide staining results.


  • Can be used as a general laboratory reagent in staining processes or for reconstitutuion of standards and controls
  • 5 gallon cube includes a spigot for easy dispensing
  • Heat sealed caps on 1 gallon bottle ensures product integrity


CAP Type I, Type II, CLSI, USP, ASTM III Type C


Current process includes a combination of cold process technologies including reverse osmosis, ion exchange resin processes, UV sterilization, and micro-filtration.
Latex Free: This product contains no latex

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Resistivity ≥10 MΩ/cm
Silicates (soluble) < 50 µg/L
Sodium ion < 10.0 µg/L
Chloride ion < 10.0 µg/L
TOC < 50 µg/L
Bacterial count < 10 CFU/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR