Matchbook™ Nitrocef
Supplier: Hardy Diagnostics
Matchbook™
Z108 Z108
CA76169-002EA 223.43 CAD
76169-692 CA76169-002
Matchbook™ Nitrocef
Microbiology Test Systems Microorganism Tests
This is a chromogenic nitrocefin test for beta lactamase, on paper strip.
  • Easy-to-use match book format
  • Easier and faster than a disk
The new Nitrocef MatchBook™ is a rapid test that allows you to quickly pick your colonies and perform the test at the same time. The rapid Nitrocef test consists of a paper match-stick that is impregnated with the Nitrocefin reagent, which is a chromogenic cephalosporin. This rapid test can yield clinically relevant information earlier than an MIC test. A positive ß-lactamase test can predict resistance to such antibiotics as penicillin, ampicillin, amoxicillin, and other ß-lactamase containing drugs for the following organisms: Neisseria gonorrhoeae, Haemophilus spp., Moraxella (Branhamella) catarrhalis, Staphylococcus spp., Enterococcus spp., Anaerobes (such as some species of Bacteroides, Porphyromonas, Clostridium, Fusobacterium, Prevotella).
The rapid Nitrocef MatchBook™ can be used for all of the above organisms with results in 5 minutes or less, with the exception of some Staphylococcus spp. that may take up to one hour. The ß-lactamase test is preferred to disk diffusion and agar dilution for the detection of plasmid-mediated beta-lactamase resistance in gonococci. Now contains a larger reaction pad.