Innoculating Needle
Supplier: Hardy Diagnostics
HSTND HSTND
CA76168-834EA 382.51 CAD
CA76168-834 76169-774
Innoculating Needle
Spreaders Inoculating Loops and Needles
Hexagonal needle shaft improves grip and is free of lubricants, oils, and electrostatic charges.
Color coded for easy product identification.
Packaging: Packaged in rectangular, plastic tubes of 50 needles with lid that allows placement on clean table without rolling; 1000 needles per box.