Sf21 Cells
Supplier: EXPRESSION SYSTEMS LLC
94-003F 94-003S
77705-754EA 837.56 CAD
77705-754 77705-756
Sf21 Cells
Cells
Expression Systems offers insect cell lines adapted to serum-free suspension culture in either ESF 921 or ESF AF.
  • Recombinant protein expression from a variety of expression platforms
  • Good growth in adherent or suspension culture
  • Quality and performance testing
The Sf21 insect cell line (IPLB-Sf-21-AE) is derived from ovarian cells of Spodoptera frugiperda.  Sf21 cells are adapted for suspension culture in ESF 921 or ESF AF and are available as a frozen vial or suspension culture.
The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.
Culture suspension shipments immediately, liquid nitrogen (vapor phase) for frozen vials.
Delivery information: Priority overnight.