Tni Cells
Supplier: EXPRESSION SYSTEMS LLC
Expression Systems offers insect cell lines adapted to serum-free suspension culture in either ESF 921 or ESF AF.
- Recombinant protein expression from a variety of expression platforms
- Can express 5 to 10× as much protein as Spodoptera cell lines
- Good growth in adherent or suspension culture
- Quality and performance testing
The Tni insect cell line is derived from ovarian cells isolated from Trichoplusia ni. Tni cells are adapted for suspension culture in ESF 921 or ESF AF and are available as a frozen vial or suspension culture.
The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.
Culture suspension shipments immediately, liquid nitrogen (vapor phase) for frozen vials.
Delivery information: Priority overnight.