Sf9 Cells
Supplier: EXPRESSION SYSTEMS LLC
94-001F 94-001S 94-006F 94-006S
77705-746EA 837.56 CAD
77705-746 77705-748 77705-760 77705-762
Sf9 Cells
Cells
Expression Systems offers insect cell lines adapted to serum-free suspension culture in either ESF 921 or ESF AF.
  • Recombinant protein expression from a variety of expression platforms
  • Good growth in adherent or suspension culture
  • Small, regular size that generates even monolayers and plaques
  • Quality and performance testing
The Sf9 insect cell line is a clonal isolate derived from the parental Spodoptera frugiperda cell line IPLB-Sf-21-AE. Sf9 cells are adapted for suspension culture in ESF 921 or ESF AF and are available as a frozen vial or suspension culture. Sf9 cells are commonly used to produce recombinant baculoviral stocks and to produce recombinant proteins.
The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.
Culture suspension shipments immediately, liquid nitrogen (vapor phase) for frozen vials.
Delivery information: Priority overnight.