Accessories for PIPETMAN® L Single Channel Pipettor, Gilson
Supplier: GILSON, INC.
F161485 F1614851 F161486 F161499 F161241
76178-118PK 42.55 CAD
76178-118 76178-120 76178-122 76178-502 76178-504 76178-506 76178-508 76178-566 76178-568 76178-570 76514-172
Accessories for PIPETMAN® L Single Channel Pipettor, Gilson
Pipettes Micropipettes
Comfort handles, replacement filters and color clips for use with PIPETMAN® pipettors.
Comfort Handles:
With a large, more comfortable finger hook and ergonomically designed handle to fit better in your hand, PIPETMAN Comfort Handle offers a better grip to support your daily pipetting activity and aids in reducing the risk of repetitive strain injuries (RSI). Labels sold seperately.
Replacement Filters:
Replacement Polyethylene filters for P5000 & P10ML Gilson pipettes.
Color Clips:
The color-coded, markable Coloris™ Identification Clips provide an easy way to clearly mark and identify PIPETMAN® pipettes, minimizing mix-ups. The Coloris clips easily snap into place without damaging or marring the pipette.