Accessories for FastPrep-96™, MP Biomedicals
Supplier: MP Biomedicals
116010560 116010560
CA76045-814EA 10663.79 CAD
CA76045-814 76020-010
Accessories for FastPrep-96™, MP Biomedicals
Homogenizers Benchtop Homogenizers
Medium-scale adapter for the FastPrep-96™ that holds 20×5 ml lysing matrix tubes.
  • Large sample volume adapter
  • Ideally suited for DNA and RNA isolation
  • Enzyme isolation
  • Protein production
  • Natural products isolation
  • Food preparation for quality analysis
  • Biopharma manufacturing
  • Forensic applications
With the FastPrep-96 Instrument, TeenFlex adapter and optimized Lysing Matrices, consistent reproducible results are obtained.
Frozen samples in Lysing Matrix tubes loaded in the adapter and stored at –20 °C or –80 °C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes. A wide range of disposable 15 ml lysing matrix tubes ensure thorough homogenization of any sample type in seconds.