Cooling or Heating Coils, Stainless Steel, PFA Coated, Chemglass
Supplier: Chemglass
CG-1966-07 CG-1966-17 CG-1966-27 CG-1972-07 CG-1972-17
47733-908EA 2045.89 CAD
47733-908 47733-918 47733-926 47734-010 47734-016
Cooling or Heating Coils, Stainless Steel, PFA Coated, Chemglass
Heating and Cooling Coils
The cooling and heating coil is made from PFA coated stainless steel and is designed to cool or heat the contents of unjacketed cylindrical reaction vessels.
  • Greem PFA coating provides complete coverage
  • Electrically tested
  • Excellent heat transfer
Design avoids problems normally associated with heat-shrink PTFE tubing on copper tubing.