Spectrometer Cuvettes, UV-Transparent, Chemglass
Supplier: Chemglass
CLS-1377-001 CLS-1377-003 CLS-1377-005 CLS-1377-101 CLS-1377-103 CLS-1377-105
30622-772EA 92.25 CAD
30622-772 30622-774 30622-776 30622-778 30622-780 30622-782
Spectrometer Cuvettes, UV-Transparent, Chemglass
The Spectrometer UV-transparent cuvettes are disposable, alternative to quartz cuvettes in DNA, RNA and protein analysis as well as many other applications between 220 nm and 900 nm are available in macro, semi-micro and ultra-micro sizes. These cuvettes eliminate the tedious and time-wasting cuvette washing and the risk of sample carry-over associated with reusable quartz cuvettes. Process QC applications avoid the need for qualified SOP’s for cuvette cleaning.
  • It has recessed windows to help protect against scratching
  • Ultra-micro UV-Cuvettes are available with 15 mm window heights that matches two standard beam heights
  • Fits most commercial spectrophotometers and photometers
The proprietary plastic material offers chemical compatibility superior to that of conventional plastic cuvettes of acrylic or polystyrene, and may be used with most polar organic solvents
Individually wrapped Ultra-micro UV-Cuvettes are available with DNA-, DNase- and RNase-Free, for use with critical samples.
They are grouped by mold cavity number to maximize consistency between cuvettes and reduce variation in extinction coefficient.