Large-Digit Triple Display Thermometers, Thermco
Supplier: Thermco
ACC895FREV ACC895INCV ACC895REFV
10025-458EA 191.04 CAD
10025-458 10025-460 10025-462
Large-Digit Triple Display Thermometers, Thermco
Thermometers Digital Thermometers
Triple display digital thermometer simultaneously shows the current, minimum, and maximum temperatures and updates continuously.
  • NIST Compliant®
  • Each thermometer is individually serialized
  • °C and °F switchable
The Triple Display Digital Thermometer simultaneously shows the current, minimum, and maximum temperatures and updates continuously.
An audible alarm will sound when the temperature rises above or goes below the set temperatures. Thermometer probe is housed in an unbreakable plastic bottle or vial filled with a Bio-Safe glycol/water or sand buffering media which maintains the thermometer reading when the instrument door is opened. The thermometer and the bottle housing the probe have dual magnets allowing attachment to doors or walls of the instrument.
The minimum/maximum feature monitors critical temperature for any period of time, such as overnight or weekend. Reset buttons on front panel.
Certifications: Each thermometer is certified and traceable to NIST.
Delivery information: Supplied with a flip-open stand, wall mounting key hole, AAA alkaline battery.