Semi-Micro Black Cuvettes, with PTFE Stopper, Firefly
Supplier: FIREFLYSCI, INC.
9MG5 9MG10 9MG20 9MG40 9MG50 9MG100 9MUV5 9MUV10 9MUV20 9MUV40 9MUV50 9MUV100 9MIR5 9MIR10 9MIR20 9MIR40 9MIR50 9MIR100
76015-292EA 247.55 CAD
76015-292 76015-294 76015-296 76015-298 76015-300 76015-302 76015-304 76015-306 76015-308 76015-310 76015-312 76015-314 76015-316 76015-318 76015-320 76015-322 76015-324 76015-326
Semi-Micro Black Cuvettes, with PTFE Stopper, Firefly
Cuvettes Spectrophotometer Cuvettes
Cuvettes are made with different materials like Optical Glass, UV quartz, IR quartz ensuring high quality optical polishing with no defects.
  • Highly accurate results
  • Fire-fused to ensure that cuvettes are robust and durable
  • Contains black walls to block out stray light
  • Base thickness is 3 mm
  • Two windows optically polished