VWR® Electronic Pipettors, Single Channel, Variable Volume
Supplier: VWR International
10827-884 10827-918 10827-920 10827-922 10861-712 10861-714 10861-716 10861-718 76035-944
10827-884EA 1250.74 CAD
10827-884 10827-918 10827-920 10827-922 10861-712 10861-714 10861-716 10861-718 76035-944
VWR® Electronic Pipettors, Single Channel, Variable Volume
Pipettes Micropipettes
VWR® electronic pipettors require virtually no force to aspirate and dispense samples, and they significantly reduce RSI (Repetitive Strain Injury) associated with routine work.
  • Ease-of-use: Only two buttons and five pipetting modes, including the CUSTOM Mode which creates pipetting protocols easily and rapidly
  • Advanced Ergonomics: Lightweight and perfectly balanced, it requires virtually no force to significantly reduce RSI
  • Fully Motorized Piston Drive: For an even and regular sample delivery across each channel
  • Large Volume Range: Five single channels (
More Product Information