Filter Funnels, Disposable, Chemglass
Supplier: Chemglass
OP-6603-C10 OP-6603-C12 OP-6603-C13
30618-008CS 344.87 CAD
30618-008 30618-010 30618-012
Filter Funnels, Disposable, Chemglass
Funnels Filtration Funnels Filter Funnels
The disposable filter funnel system for solid-liquid separation consists of a polypropylene filter funnel with 500 mg diatomaceous earth or polyethylene fritted disc, style glass vacuum adapter, and a flask or vial
The vacuum adapters are specially designed so that the vacuum can be controlled using a PTFE stopcock to prevent the filtrates from being sucked into the vacuum line. Also, the disposable filter funnels have a 80 mm long stem so that the glass vacuum adapter is not contaminated during the filtration process.
Order Now
SPECIFICATIONS
Environmentally Preferable