Rotary Evaporator Bump Traps, Fritted, Chemglass
Supplier: Chemglass
CG-1323-20 CG-1323-21 CG-1323-24
80068-124EA 313.45 CAD
80068-124 80068-126 80068-130
Rotary Evaporator Bump Traps, Fritted, Chemglass
Vacuum Traps
Rotary evaporator Bump Traps are designed with an extra coarse fritted disc sealed between the lower inner and top outer joint to prevent contents of the flask from entering into the condenser in the event of bumping.
The top outer [ST] joint on all traps is 24/40.