Foam 140® Alkaline Process and Research Cleaners
Supplier: Steris
Foam 140®
1D1405 1D1402 1D1402 1D1405
CA14004-200EA 2179.74 CAD
CA14004-200 14004-202 CA14004-202 14004-200
Foam 140® Alkaline Process and Research Cleaners
Cleaning Agents Detergents and Surface Cleansers
Foam 140® Alkaline detergent is a proprietary blend of surfactants, solvents and other performance-enhancing ingredients that provide multiple cleaning mechanisms. When applied with foam spray equipment, it generates a clinging foam to increase contact time and reduce labor costs.
  • Productivity - Reduces cleaning labor by increasing detergent contact time with heavy clinging foam when used with a foam applicator
  • Efficacy - Effectively cleans fats, oils, and other organic soils with a blend of alkalinity, surfactants and other performance-enhancing ingredients
  • Cost Savings - Reduces water consumption and cleaning time due to superior rinsing characteristics
  • Easy to Validate - Facilitates cleaning validation with a support documentation including non-specific analytical methods, and toxicity reports
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility
Foam 140® Alkaline process and research cleaner is specially formulated to deliver effective foam cleaning in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent..