Fluorescence Cuvette, Firefly
Supplier: FIREFLYSCI, INC.
52UV10X1 52UV10X2 52UV10X3 52UV10X4 52UV10X5 52IR10X1 52IR10X2 52IR10X3 52IR10X4 52IR10X5
76016-230EA 631.87 CAD
76016-230 76016-232 76016-234 76016-236 76016-238 76016-240 76016-242 76016-244 76016-246 76016-248
Fluorescence Cuvette, Firefly
Cuvettes Spectrophotometer Cuvettes
Cuvettes are made with different materials like UV quartz, IR quartz ensuring high quality optical polishing with no defects.
  • Highly accurate results
  • Fire-fused to ensure that cuvettes are robust and durable
  • Base thickness with 9 mm
  • Four windows and base optically polished