WHEATON® Headspace Screw Thread Vials and Closures, DWK LifeSciences
Supplier: DWK Life Sciences (KIMBLE)
W225284 W225286 W225285 W225287
89495-264CS 85.86 CAD
89495-264 89495-266 89495-268 89495-270
WHEATON® Headspace Screw Thread Vials and Closures, DWK LifeSciences
Vials
Headspace vials are manufactured from clear Type I borosilicate glass.
  • Clear or amber color
  • Screw thread finish eliminates the need for crimping tools
  • Accepts 18 mm screw thread closures with septa
Ordering information: Caps not included (sold separately).