VWR® Ultrasonic Homogenizers
Supplier: VWR
VWR®
76193-588 76193-590
76193-590EA 8928.94 CAD
76193-590 76193-588
VWR® Ultrasonic Homogenizers
Homogenizers Ultrasonic Cell Disruptors
VWR® Ultrasonic Homogenizers are next generation sonicators designed to simplify the process of particle size reduction and cell lysis. Designed for research labs that require stringent parameter control, reproducibility, and programmability, the VWR® Ultrasonic Homogenizers feature an intuitive touch screen interface.
  • Variable power up to 200 W
  • Programmable touch screen interface with preset programs
  • Control amplitude, energy, time, and operate in pulse or constant modes
  • Monitor processing including run time, wattage, energy, cycles, and amplitude
VWR® Ultrasonic Homogenizers are available in 50 watt or 200 watt formats
The touch screen interface supports multiple run modes, including pulse and constant operation with full amplitude control. Easily create programs that process samples based on total energy or a defined on and off time. These devices can be used for many applications, including emulsion creation, nucleic acid shearing, ChIP-Seq, nanoparticle production, protein extraction, mixing, homogenizing, or cell lysis.
The optional sound abatement chamber reduces the noise associated with sonicating. The large interior chamber easily accommodates water baths for applications requiring cooling or hot plots. The back panel of the sound abatement chamber includes six ports to allow cooling or electrical lines to be passed through the enclosure. Sealed interior walls ensure easy cleaning.
Stand assembly and additional horns are sold separately.
Certifications: CE compliant and TUV certified.
Accessories information: Accessories not included.
Ordering information: Unit supplied with power supply, transducer, horn, power cord, and user manual. The 50 watt unit includes power supply, handheld transducer, and 1/8" tip". The 200 watt unit includes power supply, transducer and 3/8" tip.