MICROMAN® Capillary Pistons, Gilson®
Supplier: GILSON, INC.
F148412 F148312 F148012 F148013 F148414 F148314 F148014 F148560
76178-124PK 237.52 CAD
76178-124 76178-126 76178-132 76178-134 76178-140 76178-144 76178-146 76178-162 76178-166
MICROMAN® Capillary Pistons, Gilson®
Pipette Tips
Designed to fit the Gilson MICROMAN™ positive displacement pipette, Capillary Piston Tips are ideal for difficult-to-pipette samples including; viscous, volatile, dense, hot or cold, or hazardous liquids. The tips work like a syringe providing protection against pipette, sample, and user contamination, and produce accurate, reproducible, and precise results. Made of pure polypropylene, their slim fit allows you to use even the narrowest and deepest test tubes.
  • Accurate and precise pipetting
  • Protection against contamination
  • Convenient
Using CapillaryMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable