VWR® TraceClean® Boston Round Glass Jars, Narrow Mouth, Clear
Supplier: VWR International
89126-548 89126-550 89126-552 89126-562 89126-656
89126-548CS 122.95 CAD
89126-548 89126-550 89126-552 89126-562 89126-656
VWR® TraceClean® Boston Round Glass Jars, Narrow Mouth, Clear
Bottles Glass Bottles
TraceClean® glass jars are used primarily for the collection of aqueous media for testing semi-volatiles, pesticides, PCBs, diesel range organics, TPH, metals, and phenolics.
Quality Assured® with VWR® Logo Sample ID Label products, a lot certificate of analysis is generated with the analytical results obtained including the concentration reporting limits and units of measurement. This certificate of analysis is included in each product case. Please refer to the applicable Certificate of Analysis for certified analytes and corresponding reporting limits.
Secondary uses include containment and archiving.
Certifications: QAL.
Ordering information: Container measurements and capacity 'contain' volumes may vary.