Airfree® Schlenk Adapter, Gas inlet, Chemglass
Supplier: Chemglass
AF-0505-10 AF-0505-11 AF-0505-12 AF-0505-13
80064-494EA 212.48 CAD
80064-494 80064-496 80064-498 80064-500
Airfree® Schlenk Adapter, Gas inlet, Chemglass
Adapters Adapters, Joints with Tube Connectors
This gas inlet adapter features a 2 mm glass stopcock having a constant pressure retension device.
  • With inner or outer standard taper joint
  • 9 mm O.D. top arm
  • With medium wall tubing
Overall height of the 14/20 size is approx. 95 mm while the 24/40 is approx. 120 mm.