Chemical Oxygen Demand (COD) Vials
Supplier: CHEMETRICS INC CA
K-7360S K-7355 CACXK-7366 K-7375
CACXK-7360EA 104 CAD
CACXK-7360 CACXK7355 CAK-7366 CACXK7375
Chemical Oxygen Demand (COD) Vials
Environmental Test Kits
COD vials are used to measure the chemical oxygen demand of influent and effluent of wastewater treatrement facilities. Requires a digestor block and photometer/spectrophotometer.
  • Leakproof reagent vials
  • Fast safe and easy COD determination
  • Works in our COD Photometer or any spectrophotometer accepting a 16 mm round cell
A fresh reagent ampoule blank must be prepared for each series of tests, therefore the number of samples that can be tested with each kit will vary.
Ordering information: COD standards are available in two ranges to run at known concentrations to verify your tests. Single Analyte Meters (SAMs) are available for photometric analysis of the COD vials.
Delivery information: Kits come in a cardboard box and contain everything needed to perform the tests (except distilled water) and instruction book.
Caution: These kits require the use of a Digestor Block along with a CHEMetrics Photometer, a COD Photometer, or a spectrophotometer capable of accepting a 16 mm round cell. All these instruments are sold separately. Unless indicated as mercury-free, the kits contain mercury. Dispose according to local, state or federal laws.
Order Now
SPECIFICATIONS
Environmentally Preferable