Nalgene® Wash Bottles, Right-to-Understand, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2428-0504 2428-0503 2428-0502 2428-0506 2428-0507
10799-119CS 415.86 CAD
10799-119 10799-049 10799-053 10799-121 10799-055
Nalgene® Wash Bottles, Right-to-Understand, Thermo Scientific
Bottles Wash Bottles
Prevent chemical accidents in the lab with Right-to-Understand LDPE, translucent wash bottles.
  • Self-venting wash bottles eliminates the risk of bottles self-dispensing
  • Good chemical resistance
  • Good temperature resistance
Bottles are pre-labeled with chemical identification and safe handling information following the 100% Globally Harmonized System (GHS) specifications as applicable to secondary laboratory containers. Wash bottles are ready to use when you transfer common laboratory chemicals from containers.