Natural Rubber Folding Skirt Stoppers, Saint-Gobain
Supplier: Saint-Gobain
DX407010-50 DX407007-100
66111-342PK 98.31 CAD
66111-342 66111-340
Natural Rubber Folding Skirt Stoppers, Saint-Gobain
Stoppers Rubber Stoppers
  • Ideal for chemical transfer
  • Temperature resistant from –4°F (–20°C) to 212°F (100°C)
The stoppers are ideal for securing glass, metal and plastic containers. Upper hollow plug section can be perforated to extract contents without removing the stopper and remains watertight even after several perforations.
The lower section grips the outside of the container and the upper hollow plug features annular serrations to provide a tight seal at the rim.