Magid® Comfort Flex® R843 Latex Gloves, Magid Glove
Supplier: Magid Glove
R843-10 R843-9
32893-152CS 1251.17 CAD
32893-152 32893-154
Magid® Comfort Flex® R843 Latex Gloves, Magid Glove
Gloves
Magid® R843 Latex, water-proof gloves provide premium protection and excellent grip in either wet or dry conditions.
  • Imprinted with hand designation for easy ID and sorting
  • Embossed palm for ideal grip
  • Rolled cuff
Ideal for handling wet materials like fisheries, canning, water and waste treatment.