PolarSafe® 100-Place EPS Microtube Racks, Argos Technologies
Supplier: Argos Technologies
PolarSafe™
6755-02 6755-01 6755-01
CA76186-144CS 1935.36 CAD
CA76186-144 CA76186-142 76196-712
PolarSafe® 100-Place EPS Microtube Racks, Argos Technologies
Boxes Cryoboxes
Secure, stackable freezer storage.
  • Accommodates microtubes and cryovials up to 2.0 mL
  • Snug-fitting lid remains in place during handling
  • Grippable indentations allow easy removal from freezer
  • Alphanumeric index in base helps locate tubes
  • Store racks safely with the stacking system
Ordering information: Two different packaging sizes are available. Choose from a pack of 10 racks, or case of 100 racks.