OK ® Sterilizer Bags, Propper Manufacturing
Supplier: Propper
O.K.™
02101100 02100200 02100300
87001-936CS 173.69 CAD
87001-936 76022-448 76022-446
OK ® Sterilizer Bags, Propper Manufacturing
Bags Sterilization Pouches
Medical grade paper bags to keep small instruments and implants safe through steam sterilization cycles.
  • All-paper, high quality gusseted sterilization bags
  • Ideal alternative to pouches or to contain small instruments or implants inside a container
  • Features pre-printed process indicator
  • Single use only
Bags are easily secured by folding over the top edge several times and then sealing with autoclave tape. These bags help in maintaining integrity even in the case of wet steam. All-paper design allows for adequate air removal steam penetration and drying while simultaneously providing a sterile barrier. Pre-printed process indicator for easy visualization and confirmation of exposure to steam. All sizes feature a gusset which allows bulky objects to be placed safely in the bag.
Superior quality medical grade paper and seals ensure integrity even in the case of wet steam. The vertical seam is double-bonded for security, and marking areas allow labeling of contents. The fold and tape closure design are ideal for quick and easy use.
Certifications: AAMI ST-79:2018