VWR® Cleanroom Socks
Supplier: VWR International
76102-192 76102-194 76102-196 76102-198
76102-192CS 2429.23 CAD
76102-192 76102-194 76102-196 76102-198
VWR® Cleanroom Socks
Workwear
Cleanroom socks provide comfort and durability for limited and extended wear in cleanroom environments.
  • Cleanroom compatible
  • No particle generation
  • Launderable
Minimize the risk of particle contamination with Cleanroom Socks from VWR®
Ideal use with VWR® Cleanroom Clogs
Available in either launderable or disposable options, Cleanroom Socks provide comfort for extended wear and are cleanroom compatible.
Packaging: Socks are packaged 1 pair per bag, 200 bags per case.