Formaldehyde 20% in aqueous solution, EM Grade
Supplier: Electron Microscopy Sciences
|
Danger
|
15713-S
15713
15713-S
100496-494EA
114.2
CAD
100496-494
100504-940
CA100496-494
Formaldehyde 20% in aqueous solution, EM Grade
Formaldehyde
Paraformaldehyde - methanol free solution.
A more efficient and rapid penetrant fixative used in combination with glutaraldehyde and acrolein fixatives.
Easy to break, prescored, 10 ml ampoules sealed under inert gas, or 100 ml bottles.
Formula:
CH₂O Boiling Pt: >93 °C (>199 °F) Density: 0.9392 g/cm³ (7.838 lbs/gal.) Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 UN: 1993 ADR: 3,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...