Formaldehyde 8% in aqueous solution, EM Grade
Supplier: Electron Microscopy Sciences
|
Danger
|
Paraformaldehyde - methanol free solution.
A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives.
Since the DOT (Department of Transportation) has changed the definition of what a non dangerous solution of formaldehyde is to under 10%, we are now offering an 8% and a 4% solution. This will allow us to ship overnight as well as overseas without any hazardous charges saving our customers a good deal of money.
Packaging: Easy to break, prescored, 10 ml ampoules sealed under inert gas, or 100 ml bottles.
Formula:
CH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 ADR: 8,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...