SP Bel-Art Tall Form Graduated Pitchers, Clear TPX, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
F28982-0000 F28984-0000 F28983-0000
13197-214CS 1084.46 CAD
13197-214 13197-212 13197-210
SP Bel-Art Tall Form Graduated Pitchers, Clear TPX, Bel-Art Products, a part of SP
Beakers
Gracefully proportioned, these pitchers are ideal for general laboratory use and offer well designed spouts and large handles helpful for use with wet or dry ingredients.
  • Crystal clear Polymethylpentene (TPX®)
  • Chemical resistant and shatterproof
  • Heat resistant to 175 °C (347 °F)
  • Steam autoclavable
Metric graduations molded on outside wall.