Packaging Buckets, Burkle
Supplier: BURKLE INC
2327-0001 2327-0020 2327-0010 2327-0030 2327-0050
76193-594EA 4.98 CAD
76193-594 76193-596 76193-598 76193-600 76193-760
Packaging Buckets, Burkle
Buckets
Packaging buckets for transport, packaging, and storing of various materials
  • Polypropylene, white finish
  • Air-tight closure
  • Supplied with lid
  • Autoclavable
  • Also available with advertising print (in large quantities on request)
Suitable for the food or building material industries and non-aggressive chemicals in environmental protection, hospitals, and the household. Buckets are equipped with a strong handle and a quality closure for product safety.