Screw Top Pathology Container, Azer Scientific
Supplier: Azer Scientific
PFNBF-20-EML
95041-504CS 259.18 CAD
95041-504
Screw Top Pathology Container, Azer Scientific
Sample Containers General Purpose Containers
This 20 mL pathology container is supplied with lid.
  • Polypropylene containers with polyethylene screw caps
  • 95 kPa compliant
This non sterile pathology container is 95 kPa compliant and comes with a screw cap lid for closure. Two clicks ensure the seal is fully engaged for safe collection and transport of the specimen.