Specimen Container, with Cap, Azer Scientific
Supplier: Azer Scientific Inc.
ES143223 ES144622
10852-644CS 365.99 CAD
10852-644 10852-646
Specimen Container, with Cap, Azer Scientific
Sample Containers General Purpose Containers
This specimen container is 95 kPa compliant and comes with a cap and temperature strip.
  • 90 mL kPa compliant container
  • Supplied with cap
  • Features a temperature strip
  • Non sterile
  • DuoClick™ closure
This non sterile specimen container comes with a cap and temperature strip. It features a DuoClick™ closure ensuring safe transportation of specimen. When the cap is turned to close, it will create an audible "click" sound, assuring the user the seal is fully engaged. The second "click" ensures 95 kPa compliance for pneumatic tube transportation.