Wallcur® PRACTI-Vials
Supplier: WALLCUR, LLC.
403PV 404MV 415TV 418MV 419MT 420PV 421TV 422TV 423PV 424PV 463PV 467PV 482MV 498PV
470232-800BX 117.49 CAD
470232-800 470232-802 470232-822 470232-828 470232-830 470232-832 470232-834 470232-836 470232-838 470232-840 470232-914 470232-922 470354-982 470354-942
Wallcur® PRACTI-Vials
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Have A Variety On Hand For Skills Training
  • Contains No Medication
  • Prefilled With Distilled Water
  • Clear And Tinted Vials Available
  • Safe For Use With Most Simulators
Teach your students the skills necessary for using all sizes of vials. These generically labeled vials are pre-filled with distilled water, so are safe to use with most simulators and injection trainers. Early training with generic vials will build confidence in your students as they prepare to work with specifically labeled vials latter in your nursing, EMS, or Pharm tech training program.
These Vials are great for teaching the following:
Small or large vial handling dexterity techniques
Aseptic precautions specific to specifically sized vials
Air pressure changes / air replacement technique
Multiple small dose calculations
Fractional dosage measurement
Delivery information: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use