Wallcur® PRACTI-Labeled Vials
Supplier: WALLCUR, LLC.
406HP 407TU 416HP 426HP 433MD 434AM 435DZ 437SC 439DH 440PM 441CC 442CG 443PC 444SP 446MP 447MSG 448MSR 449FM 450HM 453ZF 454PZ 456TC 457EG 458MP 459HB 461MR 462PF 465KM 466LP 468PV 471HL 472BX 474DZ 475NE 476SC 478SC 110LV 111ER 112LC 100EP 101TM 102CT 104RP 105LB 106EM 107HD 108NS 109GP 304DX 305BC 307KT 308CV 309VC 460SC 479EP 123DP 124PY 125MN 483VC 485LC 486AM 488FC 490TA 492RB 493CF 494SC 497SW 499FT
470232-806BX 138.49 CAD
470232-806 470232-808 470232-824 470232-844 470232-858 470232-860 470232-862 470232-866 470232-870 470232-872 470232-874 470232-876 470232-878 470232-880 470232-884 470232-886 470232-888 470232-890 470232-892 470232-896 470232-898 470232-902 470232-904 470232-906 470232-908 470232-910 470232-912 470232-918 470232-920 470232-924 470232-928 470232-930 470232-934 470232-936 470232-938 470232-942 470355-014 470354-950 470355-070 470354-932 470354-926 470355-034 470355-054 470354-962 470354-918 470355-010 470355-012 470355-060 470355-016 470354-924 470354-958 470354-940 470355-028 470354-910 470355-004 470355-050 470355-058 470354-984 470355-036 470354-968 470354-904 470355-064 470354-916 470354-930 470355-018 470355-066 470355-038 470355-000
Wallcur® PRACTI-Labeled Vials
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Prelabeled Vials For Convenience
  • Contains No Medication
  • Suitable For Most Simulator Applications
Each of these vials is already labeled in the same way as their real counterparts are! Available in a wide variety of simulated labels, each contains all the information a normal label does as well as additional warnings for training purposes. Vials are also bar coded making your pharma training easier for inventory management. Materials containing only Distilled water are generally suitable for simulator use. Those containing other materials are not recommended for use in simulators. All of these factors make these vials ideal for skills training for nursing, EMS and pharm tech programs.
Have students practice all the skills needed for proper drug selection, dosing and administration. These Vials help teach:
Reading and interpreting dosage requirements
Calculating commonly ordered antihistamine drug dosages
Small vial pressure changes
Small needle syringe techniques
Injection administration skills
Delivery information: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use